missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gamma-Undecanolactone 98.0+%, TCI America™
$32.72 - $202.15
Chemical Identifiers
| CAS | 104-67-6 |
|---|---|
| Molecular Formula | C11H20O2 |
| Molecular Weight (g/mol) | 184.279 |
| MDL Number | MFCD00005405 |
| InChI Key | PHXATPHONSXBIL-UHFFFAOYSA-N |
| Synonym | gamma-undecalactone, 4-undecanolide, 5-heptyldihydrofuran-2 3h-one, persicol, 1,4-undecanolide, peach aldehyde, peach lactone, 2 3h-furanone, 5-heptyldihydro, aldehyde c-14 peach, gamma-undecanolactone |
| PubChem CID | 7714 |
| IUPAC Name | 5-heptyloxolan-2-one |
| SMILES | CCCCCCCC1CCC(=O)O1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
U000325ML
|
TCI America
U000325ML |
25 mL |
Each for $32.72
|
|
|||||
|
U0003500ML
|
TCI America
U0003500ML |
500 mL |
Each for $202.15
|
|
|||||
Chemical Identifiers
| 104-67-6 | |
| 184.279 | |
| PHXATPHONSXBIL-UHFFFAOYSA-N | |
| 7714 | |
| CCCCCCCC1CCC(=O)O1 |
| C11H20O2 | |
| MFCD00005405 | |
| gamma-undecalactone, 4-undecanolide, 5-heptyldihydrofuran-2 3h-one, persicol, 1,4-undecanolide, peach aldehyde, peach lactone, 2 3h-furanone, 5-heptyldihydro, aldehyde c-14 peach, gamma-undecanolactone | |
| 5-heptyloxolan-2-one |
Specifications
| 104-67-6 | |
| 110°C | |
| MFCD00005405 | |
| gamma-undecalactone, 4-undecanolide, 5-heptyldihydrofuran-2 3h-one, persicol, 1,4-undecanolide, peach aldehyde, peach lactone, 2 3h-furanone, 5-heptyldihydro, aldehyde c-14 peach, gamma-undecanolactone | |
| CCCCCCCC1CCC(=O)O1 | |
| 184.279 | |
| 184.28 | |
| Liquid |
| Colorless | |
| C11H20O2 | |
| 25 mL | |
| PHXATPHONSXBIL-UHFFFAOYSA-N | |
| 5-heptyloxolan-2-one | |
| 7714 | |
| ≥98.0% (GC) | |
| gamma-Undecanolactone |
Safety and Handling
EINECSNumber : (9)-0137
RTECSNumber : YQ2485000
TSCA : Yes