missing translation for 'onlineSavingsMsg'
Learn More

gamma-Undecanolactone 98.0+%, TCI America™

Catalog No. U0003500ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U0003500ML 500 mL
U000325ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. U0003500ML Supplier TCI America Supplier No. U0003500ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 104-67-6
Molecular Formula C11H20O2
Molecular Weight (g/mol) 184.279
MDL Number MFCD00005405
InChI Key PHXATPHONSXBIL-UHFFFAOYSA-N
Synonym gamma-undecalactone, 4-undecanolide, 5-heptyldihydrofuran-2 3h-one, persicol, 1,4-undecanolide, peach aldehyde, peach lactone, 2 3h-furanone, 5-heptyldihydro, aldehyde c-14 peach, gamma-undecanolactone
PubChem CID 7714
IUPAC Name 5-heptyloxolan-2-one
SMILES CCCCCCCC1CCC(=O)O1

Specifications

Color Colorless
Boiling Point 110°C
Quantity 500 mL
Formula Weight 184.28
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material gamma-Undecanolactone
EINECSNumber (9)-0137
RTECSNumber YQ2485000
TSCA Yes