missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ gamma Tubulin Recombinant Rabbit Monoclonal Antibody (23GB4805)
GREENER_CHOICE

Catalog No. MA553832
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553832 100 μL
MA553833 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553832 Supplier Invitrogen™ Supplier No. MA553832
Only null left

Rabbit Recombinant Monoclonal Antibody

gamma Tubulin Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

This gene encodes a protein that interacts with thyroid hormone receptors, also known as hormone-dependent transcription factors. The gene product interacts specifically with the ligand binding domain. This gene is one of several that may play a role in early-stage non-small cell lung cancer.

Specifications

Antigen gamma Tubulin
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB4805
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TUBG1
Gene Accession No. P23258, P83887, P83888, Q8VCK3, Q9NRH3
Gene Alias 1500010O08Rik; AI451582; AI503389; AI504772; CDCBM4; centrosome marker; gamma tubulin; gamma-1-tubulin; Gamma-2-tubulin; gamma-tubulin; gamma-tubulin complex component 1; GCP-1; similar to tubulin, gamma 1; Tubg; TUBG1; tubg1.L; TUBG2; TUBGCP1; Tubgl; tubulin gamma 1; tubulin gamma 1 L homeolog; tubulin gamma 2; Tubulin gamma-1 chain; Tubulin gamma-2 chain; tubulin, gamma 1; tubulin, gamma 2; tubulin, gamma polypeptide; Unknown (protein for MGC:133882); XELAEV_18002398mg; xgam; zgc:55894
Gene Symbols TUBG1, Tubg2
Host Species Rabbit
Immunogen A synthesized peptide derived from human Tubulin gamma (200-451AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 103733, 103768, 252921, 27175, 680991, 7283
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less