missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ gamma Tubulin Monoclonal Antibody (4D11), Alexa Fluor™ 488
GREENER_CHOICE

Catalog No. PIMA1850A48
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA1850A48 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA1850A48 Supplier Invitrogen™ Supplier No. MA1850A488

Mouse Monoclonal Antibody

The MA1-850-A488 immunogen is a recombinant protein containing residues A A T R P D Y I S W G T Q E Q of human gamma Tubulin.

This gene encodes a protein that interacts with thyroid hormone receptors, also known as hormone-dependent transcription factors. The gene product interacts specifically with the ligand binding domain. This gene is one of several that may play a role in early-stage non-small cell lung cancer.

Specifications

Antigen gamma Tubulin
Applications Immunocytochemistry
Classification Monoclonal
Clone 4D11
Concentration 1.0 mg/mL
Conjugate Alexa Fluor 488
Formulation PBS with 1.0mg/mL BSA and 0.05% sodium azide
Gene TUBG1
Gene Accession No. P23258, P83887, Q8VCK3, Q9NRH3
Gene Alias 1500010O08Rik; AI451582; AI503389; AI504772; CDCBM4; centrosome marker; gamma tubulin; gamma-1-tubulin; Gamma-2-tubulin; gamma-tubulin; gamma-tubulin complex component 1; GCP-1; similar to tubulin, gamma 1; Tubg; TUBG1; tubg1.L; TUBG2; TUBGCP1; Tubgl; tubulin gamma 1; tubulin gamma 1 L homeolog; tubulin gamma 2; Tubulin gamma-1 chain; Tubulin gamma-2 chain; tubulin, gamma 1; tubulin, gamma 2; tubulin, gamma polypeptide; Unknown (protein for MGC:133882); XELAEV_18002398mg; xgam; zgc:55894
Gene Symbols TUBG1, Tubg2
Host Species Mouse
Immunogen Recombinant protein containing residues A A T R P D Y I S W G T Q E Q of human gamma Tubulin.
Purification Method Protein G
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 103733, 103768, 27175, 7283
Target Species Human, Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less