missing translation for 'onlineSavingsMsg'
Learn More

gamma-Oryzanol, TCI America™

Catalog No. p-7134222
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O017225G 25 g
O0172250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O017225G Supplier TCI America Supplier No. O017225G
Only null left

Chemical Identifiers

CAS 11042-64-1
Molecular Formula C40H58O4
Molecular Weight (g/mol) 602.90
MDL Number MFCD00867548
InChI Key FODTZLFLDFKIQH-FSVGXZBPSA-N
PubChem CID 91971120
IUPAC Name (1S,3R,6S,8R,11S,12S,15R,16R)-7,7,12,16-tetramethyl-15-[(2R)-6-methylhept-5-en-2-yl]pentacyclo[9.7.0.0¹,³.0³,⁸.0¹²,¹⁶]octadecan-6-yl (2E)-3-(4-hydroxy-3-methoxyphenyl)prop-2-enoate
SMILES COC1=CC(\C=C\C(=O)O[C@H]2CC[C@]34C[C@]33CC[C@]5(C)[C@H](CC[C@@]5(C)[C@@H]3CC[C@H]4C2(C)C)[C@H](C)CCC=C(C)C)=CC=C1O

Specifications

Color White-Yellow
Quantity 25 g
UN Number 2811
Physical Form Crystalline Powder
Chemical Name or Material gamma-Oryzanol
EINECSNumber (8)-0485
RTECSNumber RN0050000
TSCA No