missing translation for 'onlineSavingsMsg'
Learn More

gamma-Linolenic Acid 98.0+%, TCI America™

Supplier:  TCI America L0152100MG

 View more versions of this product

Catalog No. L0152100MG


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

gamma-Linolenic Acid
Yellow
MFCD00065718
gamma-linolenic acid, gamolenic acid, 6z,9z,12z-octadeca-6,9,12-trienoic acid, z,z,z-6,9,12-octadecatrienoic acid, ccris 7668, ligla, unii-78yc2max4o, acide gamolenique french, acido gamolenico spanish, acidum gamolenicum latin
CCCCC\C=C/C\C=C/C\C=C/CCCCC(O)=O
278.44
CHEBI:28661
≥98.0% (GC,T)
506-26-3
C18H30O2
100 mg
VZCCETWTMQHEPK-QNEBEIHSSA-N
(6Z,9Z,12Z)-octadeca-6,9,12-trienoic acid
5280933
278.44
Liquid

Chemical Identifiers

506-26-3
278.44
VZCCETWTMQHEPK-QNEBEIHSSA-N
5280933
(6Z,9Z,12Z)-octadeca-6,9,12-trienoic acid
C18H30O2
MFCD00065718
gamma-linolenic acid, gamolenic acid, 6z,9z,12z-octadeca-6,9,12-trienoic acid, z,z,z-6,9,12-octadecatrienoic acid, ccris 7668, ligla, unii-78yc2max4o, acide gamolenique french, acido gamolenico spanish, acidum gamolenicum latin
CHEBI:28661
CCCCC\C=C/C\C=C/C\C=C/CCCCC(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0609

TSCA : No

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions