accessibility menu, dialog, popup

UserName

CAS RN 506-26-3

CH3OHO(Z)(Z)(Z)

CAS RN 506-26-3

IUPAC Name: (6Z,9Z,12Z)-octadeca-6,9,12-trienoic acid
Synonyms: gamma linolenic acid indium salt gamma linolenic acid gamma-linolenic acid gamma linolenic acid, indium salt GLA gamolenic acid (product) dihomo-gamma-linolenate dibromo-gamma-linolenic acid gamolenic acid ammonium salt gamma linolenic acid
Molecular Weight (g/mol): 278.44
Molecular Formula: C18H30O2
InChi Key: VZCCETWTMQHEPK-QNEBEIHSSA-N
SMILES: CCCCC\C=C/C\C=C/C\C=C/CCCCC(O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results