missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals gamma-L-Glutamyl-L-glutamic acid, 98%

Catalog No. AC228812500
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC228812500 250 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC228812500 Supplier Thermo Scientific Chemicals Supplier No. 228812500
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1116-22-9 | C10H14N2O7 | 274.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1116-22-9
Molecular Formula C10H14N2O7
Molecular Weight (g/mol) 274.23
MDL Number MFCD00002630
InChI Key OWQDWQKWSLFFFR-WDSKDSINSA-L
Synonym gamma-glutamylglutamate, gamma-glutamylglutamic acid, gamma-l-glutamyl-l-glutamic acid, n-gamma-l-glutamyl-l-glutamic acid, 5-l-glutamyl-l-glutamate, unii-6018kcn7x2, s-2-s-4-amino-4-carboxybutanamido pentanedioic acid, 2s-2-4s-4-amino-4-carboxybutanamido pentanedioic acid, gamma-glu-glu, g-l-glutamyl-l-glutamic acid
PubChem CID 92865
ChEBI CHEBI:73705
IUPAC Name (2S)-2-[[(4S)-4-amino-4-carboxybutanoyl]amino]pentanedioic acid
SMILES [NH3+][C@@H](CCC(=O)N[C@@H](CCC([O-])=O)C([O-])=O)C([O-])=O

Specifications

Melting Point 179°C to 182°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula HO2CCH2CH2CH(CO2H)NHCOCH2CH2CH(NH2)CO2H
Quantity 250 mg
Specific Rotation Condition + 5.00 (20.00°C c=1,0.5N HCl)
Specific Rotation + 5
Solubility Information (5% in water) Clear colorless
Formula Weight 276.25
Percent Purity 98%
Physical Form Powder
Chemical Name or Material γ-L-Glutamyl-L-glutamic acid
Show More Show Less
EINECSNumber 214-233-2

RUO – Research Use Only