CAS RN 1116-22-9
IUPAC Name:
(2S)-2-[(4S)-4-azaniumyl-4-carboxylatobutanamido]pentanedioate
Synonyms:
(2S)-2-[(4S)-4-ammonio-4-carboxylatobutanamido]pentanedioate
Molecular Weight (g/mol):
274.23
Molecular Formula:
C10H14N2O7
InChi Key:
OWQDWQKWSLFFFR-WDSKDSINSA-L
SMILES:
[NH3+][C@@H](CCC(=O)N[C@@H](CCC([O-])=O)C([O-])=O)C([O-])=O
1
–
1
of
1
results
1
Promotions Available
Thermo Scientific Chemicals gamma-L-Glutamyl-L-glutamic acid, 98%
Thermo Scientific Chemicals gamma-L-Glutamyl-L-glutamic acid, 98%
CAS: 1116-22-9 Molecular Formula: C10H14N2O7 Molecular Weight (g/mol): 274.23 MDL Number: MFCD00002630 InChI Key: OWQDWQKWSLFFFR-WDSKDSINSA-L Synonym: gamma-glutamylglutamate,gamma-glutamylglutamic acid,gamma-l-glutamyl-l-glutamic acid,n-gamma-l-glutamyl-l-glutamic acid,5-l-glutamyl-l-glutamate,unii-6018kcn7x2,s-2-s-4-amino-4-carboxybutanamido pentanedioic acid,2s-2-4s-4-amino-4-carboxybutanamido pentanedioic acid,gamma-glu-glu,g-l-glutamyl-l-glutamic acid PubChem CID: 92865 ChEBI: CHEBI:73705 IUPAC Name: (2S)-2-[[(4S)-4-amino-4-carboxybutanoyl]amino]pentanedioic acid SMILES: [NH3+][C@@H](CCC(=O)N[C@@H](CCC([O-])=O)C([O-])=O)C([O-])=O