Learn More
gamma-Hexanolactone, 98%
CAS: 695-06-7 | C6H10O2 | 114.14 g/mol
Supplier: Thermo Scientific Chemicals A1964222
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| gamma-Hexanolactone | |
| -18°C | |
| 218°C to 219°C | |
| C6H10O2 | |
| MFCD00005401 | |
| 107260 | |
| JBFHTYHTHYHCDJ-UHFFFAOYNA-N | |
| 5-ethyloxolan-2-one | |
| 12756 | |
| 114.15 |
| 695-06-7 | |
| 1.025 | |
| 98°C (208°F) | |
| 1.439 | |
| 100 g | |
| gamma-caprolactone, gamma-hexalactone, 4-hexanolide, gamma-hexanolactone, 6-caprolactone, tonkalide, toukalide, 2 3h-furanone, 5-ethyldihydro, 4-ethyl-4-butanolide, 5-ethyldihydrofuran-2 3h-one | |
| CCC1CCC(=O)O1 | |
| 114.14 | |
| CHEBI:85235 | |
| 98% |
Chemical Identifiers
| 695-06-7 | |
| 114.14 | |
| JBFHTYHTHYHCDJ-UHFFFAOYNA-N | |
| 12756 | |
| 5-ethyloxolan-2-one |
| C6H10O2 | |
| MFCD00005401 | |
| gamma-caprolactone, gamma-hexalactone, 4-hexanolide, gamma-hexanolactone, 6-caprolactone, tonkalide, toukalide, 2 3h-furanone, 5-ethyldihydro, 4-ethyl-4-butanolide, 5-ethyldihydrofuran-2 3h-one | |
| CHEBI:85235 | |
| CCC1CCC(=O)O1 |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
GHS P Statement
P280-P264-P305+P351+P338-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
Wash thoroughly after handling.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF eye irritation persists: Get medical advice/attention.
Warning
EINECSNumber : 211-778-8
RTECSNumber : LU4220000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only