accessibility menu, dialog, popup

UserName

CAS RN 695-06-7

IUPAC Name: 5-ethyloxolan-2-one
Synonyms: gamma-hexalactone gamma-caprolactone 4-hexanolide gamma-hexanolactone hexanolide-1,4 6-caprolactone toukalide hexa-4-olide hexan-4-olide tonkalide
Molecular Weight (g/mol): 114.14
Molecular Formula: C6H10O2
InChi Key: JBFHTYHTHYHCDJ-UHFFFAOYNA-N
SMILES: CCC1CCC(=O)O1
Boiling Point 
  • (2)
  • (1)
  • (3)
Percent Purity 
  • (1)
  • (5)
Quantity 
  • (2)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (5)
  • (2)
  • (1)
  • (3)
  • (1)
  • (5)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)