missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gamma-Heptanolactone 98.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Boiling Point | 236°C |
| Quantity | 25 mL |
| Formula Weight | 128.17 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Liquid |
| Chemical Name or Material | gamma-Heptanolactone |
Safety and Handling
| EINECSNumber | (9)-0137 |
| RTECSNumber | LU3697000 |
| TSCA | Yes |