accessibility menu, dialog, popup

UserName

CAS RN 105-21-5

IUPAC Name: 5-propyloxolan-2-one
Synonyms: gamma-heptalactone
Molecular Weight (g/mol): 128.17
Molecular Formula: C7H12O2
InChi Key: VLSVVMPLPMNWBH-UHFFFAOYNA-N
SMILES: CCCC1CCC(=O)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results