missing translation for 'onlineSavingsMsg'
Learn More

gamma-Decanolactone, 98%

CAS: 706-14-9 | C10H18O2 | 170.25 g/mol

Supplier:  Thermo Scientific Chemicals A1973214

 View more versions of this product

Catalog No. AAA1973214

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

gamma-Decanolactone
0.948
146°C (294°F)
1.5
25 g
gamma-decalactone, 4-decanolide, gamma-decanolactone, 2 3h-furanone, 5-hexyldihydro, 5-hexyldihydrofuran-2 3h-one, decan-4-olide, 5-hexyldihydro-2 3h-furanone, decanolide-1,4, 2-decalactone, 4-hexyl-4-butanolide
CCCCCCC1CCC(=O)O1
170.25
170.25
706-14-9
150°C to 153°C (15 mmHg)
C10H18O2
MFCD00005404
117547
IFYYFLINQYPWGJ-UHFFFAOYNA-N
5-hexyloxolan-2-one
12813
98%

Chemical Identifiers

706-14-9
170.25
IFYYFLINQYPWGJ-UHFFFAOYNA-N
12813
CCCCCCC1CCC(=O)O1
C10H18O2
MFCD00005404
gamma-decalactone, 4-decanolide, gamma-decanolactone, 2 3h-furanone, 5-hexyldihydro, 5-hexyldihydrofuran-2 3h-one, decan-4-olide, 5-hexyldihydro-2 3h-furanone, decanolide-1,4, 2-decalactone, 4-hexyl-4-butanolide
5-hexyloxolan-2-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 211-892-8

RTECSNumber : LU4600000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only