missing translation for 'onlineSavingsMsg'
Learn More

gamma-Decanolactone, 98%

Catalog No. p-7024664
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
5 g
Catalog No. Quantity
AAA1973206 5 g
AAA1973214 25 g
AAA1973222 100 g
3 options

Catalog No. AAA1973206

Supplier: Thermo Scientific Chemicals A1973206

Only null left

CAS: 706-14-9 | C10H18O2 | 170.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 706-14-9
Molecular Formula C10H18O2
Molecular Weight (g/mol) 170.25
MDL Number MFCD00005404
InChI Key IFYYFLINQYPWGJ-UHFFFAOYNA-N
Synonym gamma-decalactone, 4-decanolide, gamma-decanolactone, 2 3h-furanone, 5-hexyldihydro, 5-hexyldihydrofuran-2 3h-one, decan-4-olide, 5-hexyldihydro-2 3h-furanone, decanolide-1,4, 2-decalactone, 4-hexyl-4-butanolide
PubChem CID 12813
IUPAC Name 5-hexyloxolan-2-one
SMILES CCCCCCC1CCC(=O)O1

Specifications

Density 0.948
Boiling Point 150°C to 153°C (15 mmHg)
Flash Point 146°C (294°F)
Refractive Index 1.5
Quantity 5 g
Beilstein 117547
Formula Weight 170.25
Percent Purity 98%
Chemical Name or Material gamma-Decanolactone
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 211-892-8
RTECSNumber LU4600000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only