missing translation for 'onlineSavingsMsg'
Learn More
Learn More
gamma-Cyclodextrin 99.0+%, TCI America™
Supplier: TCI America C086925G
| Quantity | 25 g |
|---|
Chemical Identifiers
| 17465-86-0 | |
| 1297.13 | |
| GDSRMADSINPKSL-UHFFFAOYNA-N | |
| 101136809 | |
| OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC9C(CO)OC(OC1C(O)C2O)C(O)C9O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O |
| C48H80O40 | |
| MFCD00009595,MFCD00149574 | |
| cyclooctaamylose, schardinger gamma-dextrin | |
| 5,10,15,20,25,30,35,40-octakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34,37,39-hexadecaoxanonacyclo[36.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹.2³³,³⁶]hexapentacontane-41,42,43,44,45,46,47,48,49,50,51,52,53,54,55,56-hexadecol |
Specifications
| gamma-Cyclodextrin | |
| White | |
| 25 g | |
| cyclooctaamylose, schardinger gamma-dextrin | |
| OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC9C(CO)OC(OC1C(O)C2O)C(O)C9O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O | |
| 1297.13 | |
| 1297.13 | |
| Crystalline Powder |
| 17465-86-0 | |
| C48H80O40 | |
| MFCD00009595,MFCD00149574 | |
| GDSRMADSINPKSL-UHFFFAOYNA-N | |
| 5,10,15,20,25,30,35,40-octakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34,37,39-hexadecaoxanonacyclo[36.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹.2³³,³⁶]hexapentacontane-41,42,43,44,45,46,47,48,49,50,51,52,53,54,55,56-hexadecol | |
| 101136809 | |
| ≥99.0% (HPLC) |
Safety and Handling
EINECSNumber : (8)-0101
RTECSNumber : GU2293080
TSCA : Yes