missing translation for 'onlineSavingsMsg'
Learn More

gamma-Cyclodextrin 99.0+%, TCI America™

Catalog No. p-7124655
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C08695G 5 g
C086925G 25 g
C0869100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C08695G Supplier TCI America Supplier No. C08695G

Chemical Identifiers

CAS 17465-86-0
Molecular Formula C48H80O40
Molecular Weight (g/mol) 1297.13
MDL Number MFCD00009595,MFCD00149574
InChI Key GDSRMADSINPKSL-UHFFFAOYNA-N
Synonym cyclooctaamylose, schardinger gamma-dextrin
PubChem CID 101136809
IUPAC Name 5,10,15,20,25,30,35,40-octakis(hydroxymethyl)-2,4,7,9,12,14,17,19,22,24,27,29,32,34,37,39-hexadecaoxanonacyclo[36.2.2.2³,⁶.2⁸,¹¹.2¹³,¹⁶.2¹⁸,²¹.2²³,²⁶.2²⁸,³¹.2³³,³⁶]hexapentacontane-41,42,43,44,45,46,47,48,49,50,51,52,53,54,55,56-hexadecol
SMILES OCC1OC2OC3C(CO)OC(OC4C(CO)OC(OC5C(CO)OC(OC6C(CO)OC(OC7C(CO)OC(OC8C(CO)OC(OC9C(CO)OC(OC1C(O)C2O)C(O)C9O)C(O)C8O)C(O)C7O)C(O)C6O)C(O)C5O)C(O)C4O)C(O)C3O

Specifications

Color White
Quantity 5 g
Formula Weight 1297.13
Percent Purity ≥99.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material gamma-Cyclodextrin
EINECSNumber (8)-0101
RTECSNumber GU2293080
TSCA Yes