missing translation for 'onlineSavingsMsg'
Learn More

gamma-Crotonolactone 93.0+%, TCI America™

Catalog No. C10785ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C10785ML 5 mL
C107825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C10785ML Supplier TCI America Supplier No. C10785ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 497-23-4
Molecular Formula C4H4O2
Molecular Weight (g/mol) 84.07
MDL Number MFCD00005376
InChI Key VIHAEDVKXSOUAT-UHFFFAOYSA-N
Synonym 2 5h-furanone, furan-2 5h-one, butenolide, 2-butenolide, gamma-crotonolactone, 2-buten-4-olide, isocrotonolactone, 2-oxo-2,5-dihydrofuran, crotonolactone, 2-5h-furanone
PubChem CID 10341
ChEBI CHEBI:38118
IUPAC Name 2,5-dihydrofuran-2-one
SMILES O=C1OCC=C1

Specifications

Melting Point 5°C
Color Yellow
Boiling Point 214°C
Quantity 5 mL
Formula Weight 84.07
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material gamma-Crotonolactone
RTECSNumber LU3453000
TSCA No
Recommended Storage Refrigerator