accessibility menu, dialog, popup

UserName

CAS RN 497-23-4

IUPAC Name: 2,5-dihydrofuran-2-one
Synonyms: 2-(5H)-furanone gamma-crotonolactone 2-butenoic acid-gamma-lactone crotonolactone delta,α,β-butenolide 2-buten-4-olide gamma-crotolactone 2-butenoic acid gamma-lactone α,β-crotonolactone butenolide
Molecular Weight (g/mol): 84.07
Molecular Formula: C4H4O2
InChi Key: VIHAEDVKXSOUAT-UHFFFAOYSA-N
SMILES: O=C1OCC=C1
Boiling Point 
  • (3)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (5)
  • (3)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)