missing translation for 'onlineSavingsMsg'
Learn More

Galvinoxyl Free Radical, TCI America™

Catalog No. G00201G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G00201G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. G00201G Supplier TCI America Supplier No. G00201G
Only null left

Chemical Identifiers

CAS 2370-18-5
Molecular Formula C29H42O2
Molecular Weight (g/mol) 422.65
MDL Number MFCD00001616
InChI Key HRLDHROCDBYHAU-UHFFFAOYSA-N
IUPAC Name 2,6-di-tert-butyl-4-[(3,5-di-tert-butyl-4-hydroxyphenyl)methylidene]cyclohexa-2,5-dien-1-one
SMILES CC(C)(C)C1=CC(C=C2C=C(C(=O)C(=C2)C(C)(C)C)C(C)(C)C)=CC(=C1O)C(C)(C)C

Specifications

Color Blue-Purple
Quantity 1 g
Formula Weight 421.65
Physical Form Crystalline Powder
Chemical Name or Material Galvinoxyl Free Radical
RTECSNumber XV7978000
TSCA No
Recommended Storage Refrigerator