accessibility menu, dialog, popup

UserName

CAS RN 2370-18-5

CH3H3CCH3OH3CH3CCH3CH3H3CCH3HOH3CH3CCH3

CAS RN 2370-18-5

IUPAC Name: 2,6-di-tert-butyl-4-[(3,5-di-tert-butyl-4-hydroxyphenyl)methylidene]cyclohexa-2,5-dien-1-one
Synonyms: 2,6-di-tert-butyl-4-[(3,5-di-tert-butyl-4-hydroxyphenyl)methylidene]cyclohexa-2,5-dien-1-one
Molecular Weight (g/mol): 422.65
Molecular Formula: C29H42O2
InChi Key: HRLDHROCDBYHAU-UHFFFAOYSA-N
SMILES: CC(C)(C)C1=CC(C=C2C=C(C(=O)C(=C2)C(C)(C)C)C(C)(C)C)=CC(=C1O)C(C)(C)C
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results