Learn More
Gallium phosphide, 99.999% (metals basis)
For Use in Semiconducting Devices | CAS: 12063-98-8 | GaP | 100.70 g/mol
Supplier: Thermo Scientific Chemicals 01226618
Description
Gallium phosphide (GaP) is used in semiconductor devices, and in the manufacture of light-emitting diodes (LEDs) doped with other elements or in combination with gallium arsenide phosphide. With appropriate dopants, p-type and n-type semiconductors can be produced. It is used in optical systems.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| Gallium phosphide | |
| Orange | |
| Pieces | |
| 99.999% (Metals basis) | |
| MFCD00016109 | |
| Insoluble in water. | |
| P#[Ga] | |
| 100.70 | |
| 100.69 |
| 1465°C | |
| 12063-98-8 | |
| 50 g | |
| GaP | |
| gallium phosphide, gallium monophosphide, unii-3j421f73dv, ccris 4019, gallium phosphide, polycryst, gallium phosphide trace metals basis, gallium phosphide trace metals basis 1g, gallium phosphide, single crystal substrate , <111>, diam. x thickness 2 in. x 0.5 mm | |
| HZXMRANICFIONG-UHFFFAOYSA-N | |
| gallanylidynephosphane | |
| 82901 | |
| 4.13 |
Chemical Identifiers
| 12063-98-8 | |
| 100.70 | |
| HZXMRANICFIONG-UHFFFAOYSA-N | |
| 82901 | |
| P#[Ga] |
| GaP | |
| MFCD00016109 | |
| gallium phosphide, gallium monophosphide, unii-3j421f73dv, ccris 4019, gallium phosphide, polycryst, gallium phosphide trace metals basis, gallium phosphide trace metals basis 1g, gallium phosphide, single crystal substrate , <111>, diam. x thickness 2 in. x 0.5 mm | |
| gallanylidynephosphane |
Safety and Handling
In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. This material and/or its container must be disposed of as hazardous waste.
P261-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P501c
H319-H335
EINECSNumber : 235-057-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only