missing translation for 'onlineSavingsMsg'
Learn More

Gallium(III) sulfide, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog No. p-7051562
Change view
Click to view available options
Quantity:
0.2 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41538SS 0.2 g
AA4153806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA41538SS Supplier Thermo Scientific Chemicals Supplier No. 41538SS
Only null left

Gallium(III) sulfide is used in semiconductor materials, functional ceramic materials, fluorescent powder and photoelectric materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12024-22-5
Molecular Formula Ga2S3
Molecular Weight (g/mol) 235.63
MDL Number MFCD00016106
InChI Key BVSHTEBQPBBCFT-UHFFFAOYSA-N
IUPAC Name digallium(3+) trisulfanediide
SMILES [S--].[S--].[S--].[Ga+3].[Ga+3]

Specifications

Melting Point 1255°C
Physical Form Powder
Quantity 0.2 g
UN Number UN3260
Solubility Information It dissolves in water slowly,which is easy to dissolve in concentrated alkaline to get gallate salts. It is also dissolves in HCL and nitric acid.
Formula Weight 235.63
Percent Purity 99.999%
Odor Odorless
Sensitivity Moisture sensitive
Density 3.77 g/mL
Assay (metals basis)
Chemical Name or Material Gallium(III) sulfide
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 234-692-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only