accessibility menu, dialog, popup

UserName

SSSGaGa

CAS RN 12024-22-5

IUPAC Name: digallium(3+) trisulfanediide
Synonyms: gallium(III) sulfide
Molecular Weight (g/mol): 235.63
Molecular Formula: Ga2S3
InChi Key: BVSHTEBQPBBCFT-UHFFFAOYSA-N
SMILES: [S--].[S--].[S--].[Ga+3].[Ga+3]
Percent Purity 
  • (3)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (3)
  • (2)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)