missing translation for 'onlineSavingsMsg'
Learn More

Gallium(III) Chloride Anhydrous 98.0+%, TCI America™

Supplier:  TCI America G03595G

 View more versions of this product

Catalog No. G03595G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

201°C
78°C
13450-90-3
5 g
MFCD00011018
gallium trichloride, gallium iii chloride, trichlorogallium, gallium chloride, gallium chloride gacl3, gallium 3+ chloride, gacl3, ccris 4492, gallium iii chloride, ultra dry, trichlorogalliue
Cl[Ga](Cl)Cl
176.073
176.07
Gallium(III) Chloride Anhydrous
White
Crystalline Powder
Cl3Ga
3260
UPWPDUACHOATKO-UHFFFAOYSA-K
trichlorogallane
26010
≥98.0%

Chemical Identifiers

13450-90-3
176.073
UPWPDUACHOATKO-UHFFFAOYSA-K
26010
Cl[Ga](Cl)Cl
Cl3Ga
MFCD00011018
gallium trichloride, gallium iii chloride, trichlorogallium, gallium chloride, gallium chloride gacl3, gallium 3+ chloride, gacl3, ccris 4492, gallium iii chloride, ultra dry, trichlorogalliue
trichlorogallane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : LW9100000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions