missing translation for 'onlineSavingsMsg'
Learn More

Gallium(III) Chloride Anhydrous 98.0+%, TCI America™

Catalog No. p-7130245
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G03595G 5 g
G035925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G03595G Supplier TCI America Supplier No. G03595G
Only null left

Chemical Identifiers

CAS 13450-90-3
Molecular Formula Cl3Ga
Molecular Weight (g/mol) 176.073
MDL Number MFCD00011018
InChI Key UPWPDUACHOATKO-UHFFFAOYSA-K
Synonym gallium trichloride, gallium iii chloride, trichlorogallium, gallium chloride, gallium chloride gacl3, gallium 3+ chloride, gacl3, ccris 4492, gallium iii chloride, ultra dry, trichlorogalliue
PubChem CID 26010
IUPAC Name trichlorogallane
SMILES Cl[Ga](Cl)Cl

Specifications

Boiling Point 201°C
Melting Point 78°C
Color White
Physical Form Crystalline Powder
Quantity 5 g
UN Number 3260
Formula Weight 176.07
Percent Purity ≥98.0%
Chemical Name or Material Gallium(III) Chloride Anhydrous
RTECSNumber LW9100000
TSCA Yes