Learn More
Gallium arsenide, 99.999% (metals basis)
For Semiconductors (Transistors, Lasers, Solar Cells) | CAS: 1303-00-0 | AsGa | 144.645 g/mol
$229.49 - $4,543.89
Chemical Identifiers
| CAS | 1303-00-0 |
|---|---|
| Molecular Formula | AsGa |
| Molecular Weight (g/mol) | 144.645 |
| MDL Number | MFCD00011017 |
| InChI Key | JBRZTFJDHDCESZ-UHFFFAOYSA-N |
| Synonym | gallium arsenide, gallium arsenide gaas, gallium monoarsenide, gallium monoarsenide gaas, unii-27fc46ga44, ccris 4020, arsinogallane, gallium arsenide trace metals basis 1g, gallium arsenide, pieces trace metals basis, gallium arsenide optical window, 12.7mm 0.5in dia x 1mm 0.04in thick |
| PubChem CID | 14770 |
| IUPAC Name | gallanylidynearsane |
| SMILES | [Ga]#[As] |
Description
Semiconductors (transistors, lasers, solar cells)Gallium arsenide is used in semiconductor applications. It is also used in the manufacture of devices such as microwave frequency integrated circuits, Gunn diodes, monolithic microwave integrated circuits, infrared light-emitting diodes, laser diodes and optical windows. Further, it is used for single-crystalline thin film solar cells and multi-junction solar cells. In addition, it is used for the detection of X-rays.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1303-00-0 | |
| 144.645 | |
| JBRZTFJDHDCESZ-UHFFFAOYSA-N | |
| 14770 | |
| [Ga]#[As] |
| AsGa | |
| MFCD00011017 | |
| gallium arsenide, gallium arsenide gaas, gallium monoarsenide, gallium monoarsenide gaas, unii-27fc46ga44, ccris 4020, arsinogallane, gallium arsenide trace metals basis 1g, gallium arsenide, pieces trace metals basis, gallium arsenide optical window, 12.7mm 0.5in dia x 1mm 0.04in thick | |
| gallanylidynearsane |
Specifications
| 1238°C | |
| Gray | |
| 2 g | |
| AsGa | |
| gallium arsenide, gallium arsenide gaas, gallium monoarsenide, gallium monoarsenide gaas, unii-27fc46ga44, ccris 4020, arsinogallane, gallium arsenide trace metals basis 1g, gallium arsenide, pieces trace metals basis, gallium arsenide optical window, 12.7mm 0.5in dia x 1mm 0.04in thick | |
| JBRZTFJDHDCESZ-UHFFFAOYSA-N | |
| gallanylidynearsane | |
| 14770 | |
| Odorless | |
| Gallium arsenide |
| 1303-00-0 | |
| Polycrystalline Pieces | |
| 99.999% (Metals basis) | |
| MFCD00011017 | |
| Soluble in hydrochloric acid. Insoluble in water,ethanol,methanol and acetone. | |
| [Ga]#[As] | |
| 144.645 | |
| 144.64 | |
| 5.31 g/cm3 |
Safety and Handling
When using do not eat, drink or smoke. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment.
P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c
H301+H331-H350
DOTInformation : DOT Class: 6.1, Packing Group: II
EINECSNumber : 215-114-8
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only