missing translation for 'onlineSavingsMsg'
Learn More

Gallium arsenide, 99.999% (metals basis)

For Semiconductors (Transistors, Lasers, Solar Cells) | CAS: 1303-00-0 | AsGa | 144.645 g/mol

Supplier:  Thermo Scientific Chemicals 08845809

 View more versions of this product

Catalog No. AA8845809


Only null left
Explore available promotions

Description

Description

Semiconductors (transistors, lasers, solar cells)Gallium arsenide is used in semiconductor applications. It is also used in the manufacture of devices such as microwave frequency integrated circuits, Gunn diodes, monolithic microwave integrated circuits, infrared light-emitting diodes, laser diodes and optical windows. Further, it is used for single-crystalline thin film solar cells and multi-junction solar cells. In addition, it is used for the detection of X-rays.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Gallium arsenide
1303-00-0
Polycrystalline Pieces
99.999% (Metals basis)
MFCD00011017
Soluble in hydrochloric acid. Insoluble in water,ethanol,methanol and acetone.
[Ga]#[As]
144.645
144.64
5.31
1238°C
Gray
10 g
AsGa
gallium arsenide, gallium arsenide gaas, gallium monoarsenide, gallium monoarsenide gaas, unii-27fc46ga44, ccris 4020, arsinogallane, gallium arsenide trace metals basis 1g, gallium arsenide, pieces trace metals basis, gallium arsenide optical window, 12.7mm 0.5in dia x 1mm 0.04in thick
JBRZTFJDHDCESZ-UHFFFAOYSA-N
gallanylidynearsane
14770
Odorless

Chemical Identifiers

1303-00-0
144.645
JBRZTFJDHDCESZ-UHFFFAOYSA-N
14770
[Ga]#[As]
AsGa
MFCD00011017
gallium arsenide, gallium arsenide gaas, gallium monoarsenide, gallium monoarsenide gaas, unii-27fc46ga44, ccris 4020, arsinogallane, gallium arsenide trace metals basis 1g, gallium arsenide, pieces trace metals basis, gallium arsenide optical window, 12.7mm 0.5in dia x 1mm 0.04in thick
gallanylidynearsane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

When using do not eat, drink or smoke. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment.

P201-P202-P261-P264b-P270-P271-P281-P301+P310-P304+P340-P308+P313-P311-P330-P501c

H301+H331-H350

DOTInformation : DOT Class: 6.1, Packing Group: II

EINECSNumber : 215-114-8

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only