missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gallic Acid Hydrate 98.0+%, TCI America™
Supplier: TCI America G0011100G
Specifications
| Gallic Acid Hydrate | |
| 5995-86-8 | |
| C7H5O5 | |
| gallic acid monohydrate, 3,4,5-trihydroxybenzoic acid hydrate, galop hydrate, 3,4,5-trihydroxybenzoic acid monohydrate, gallicum acidum, benzoic acid, 3,4,5-trihydroxy-, hydrate, gallic acid, monohydrate, gallicacidmonohydrate, gallic acid, acs, acmc-1ary0 | |
| OC1=CC(=CC(O)=C1O)C([O-])=O | |
| 169.11 | |
| 170.12 | |
| 100 g |
| 253°C | |
| White-Yellow | |
| MFCD00149098 | |
| LNTHITQWFMADLM-UHFFFAOYSA-M | |
| 3,4,5-trihydroxybenzoate | |
| 24721416 | |
| ≥98.0% (HPLC,T) | |
| Crystalline Powder |
Chemical Identifiers
| 5995-86-8 | |
| 169.11 | |
| LNTHITQWFMADLM-UHFFFAOYSA-M | |
| 24721416 | |
| OC1=CC(=CC(O)=C1O)C([O-])=O |
| C7H5O5 | |
| MFCD00149098 | |
| gallic acid monohydrate, 3,4,5-trihydroxybenzoic acid hydrate, galop hydrate, 3,4,5-trihydroxybenzoic acid monohydrate, gallicum acidum, benzoic acid, 3,4,5-trihydroxy-, hydrate, gallic acid, monohydrate, gallicacidmonohydrate, gallic acid, acs, acmc-1ary0 | |
| 3,4,5-trihydroxybenzoate |
Safety and Handling
EINECSNumber : (3)-1629&(9)-1351
RTECSNumber : LW7525000
TSCA : Yes