missing translation for 'onlineSavingsMsg'
Learn More

Gallic Acid Hydrate 98.0+%, TCI America™

Catalog No. p-7130097
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G001125G 25 g
G0011100G 100 g
G0011500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. G001125G Supplier TCI America Supplier No. G001125G

Chemical Identifiers

CAS 5995-86-8
Molecular Formula C7H5O5
Molecular Weight (g/mol) 169.11
MDL Number MFCD00149098
InChI Key LNTHITQWFMADLM-UHFFFAOYSA-M
Synonym gallic acid monohydrate, 3,4,5-trihydroxybenzoic acid hydrate, galop hydrate, 3,4,5-trihydroxybenzoic acid monohydrate, gallicum acidum, benzoic acid, 3,4,5-trihydroxy-, hydrate, gallic acid, monohydrate, gallicacidmonohydrate, gallic acid, acs, acmc-1ary0
PubChem CID 24721416
IUPAC Name 3,4,5-trihydroxybenzoate
SMILES OC1=CC(=CC(O)=C1O)C([O-])=O

Specifications

Melting Point 253°C
Color White-Yellow
Formula Weight 170.12
Percent Purity ≥98.0% (HPLC,T)
Quantity 25 g
Physical Form Crystalline Powder
Chemical Name or Material Gallic Acid Hydrate
EINECSNumber (3)-1629&(9)-1351
RTECSNumber LW7525000
TSCA Yes