missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gallic Acid Standard, 500 ppm C7H6O5, Ricca Chemical
Spécifications
Spécifications
| Nom ou substance chimique | Gallic Acid Standard, 500ppm |
| Emballage | Natural Poly Bottle |
| CAS % minimum | 98.0, 0.05 |
| CAS % maximal | 100.0, 0.06 |