missing translation for 'onlineSavingsMsg'
Learn More

Gallic Acid Standard, 2000 ppm C7H6O5, Ricca Chemical

Catalog No. R3224200100
Change view
Click to view available options
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Packaging
R3224200100 Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R3224200100 Supplier Ricca Chemical Company Supplier No. R3224200100A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 5995-86-8 , 7732-18-5
Molecular Formula C7H5O5
Molecular Weight (g/mol) 169.11
MDL Number MFCD00149098
InChI Key LNTHITQWFMADLM-UHFFFAOYSA-M
PubChem CID 24721416
IUPAC Name 3,4,5-trihydroxybenzoate
SMILES OC1=CC(=CC(O)=C1O)C([O-])=O

Specifications

Packaging Natural Poly Bottle
CAS Min % 98.0, 0.22
CAS Max % 100.0, 0.23
Chemical Name or Material Gallic Acid Standard, 2000ppm