missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gallein 85.0+%, TCI America™
$84.70 - $731.56
Chemical Identifiers
| CAS | 2103-64-2 |
|---|---|
| Molecular Formula | C20H12O7 |
| Molecular Weight (g/mol) | 364.309 |
| MDL Number | MFCD00041839 |
| InChI Key | PHLYOKFVXIVOJC-UHFFFAOYSA-N |
| Synonym | gallein, alizarin violet, pyrogallolphthalein, c.i. mordant violet 25, 4,5-dihydroxyfluorescein, mordant violet 25, 3',4',5',6'-tetrahydroxy-3h-spiro isobenzofuran-1,9'-xanthen-3-one, alizarine violet, unii-8l0084u2qr, fluorescein, 4',5'-dihydroxy |
| PubChem CID | 73685 |
| IUPAC Name | 3',4',5',6'-tetrahydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one |
| SMILES | C1=CC=C2C(=C1)C(=O)OC23C4=C(C(=C(C=C4)O)O)OC5=C3C=CC(=C5O)O |
Chemical Identifiers
| 2103-64-2 | |
| 364.309 | |
| PHLYOKFVXIVOJC-UHFFFAOYSA-N | |
| 73685 | |
| C1=CC=C2C(=C1)C(=O)OC23C4=C(C(=C(C=C4)O)O)OC5=C3C=CC(=C5O)O |
| C20H12O7 | |
| MFCD00041839 | |
| gallein, alizarin violet, pyrogallolphthalein, c.i. mordant violet 25, 4,5-dihydroxyfluorescein, mordant violet 25, 3',4',5',6'-tetrahydroxy-3h-spiro isobenzofuran-1,9'-xanthen-3-one, alizarine violet, unii-8l0084u2qr, fluorescein, 4',5'-dihydroxy | |
| 3',4',5',6'-tetrahydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one |
Specifications
| 2103-64-2 | |
| C20H12O7 | |
| 1 g | |
| PHLYOKFVXIVOJC-UHFFFAOYSA-N | |
| 3',4',5',6'-tetrahydroxyspiro[2-benzofuran-3,9'-xanthene]-1-one | |
| 73685 | |
| ≥85.0% (HPLC) | |
| Gallein |
| Black | |
| MFCD00041839 | |
| gallein, alizarin violet, pyrogallolphthalein, c.i. mordant violet 25, 4,5-dihydroxyfluorescein, mordant violet 25, 3',4',5',6'-tetrahydroxy-3h-spiro isobenzofuran-1,9'-xanthen-3-one, alizarine violet, unii-8l0084u2qr, fluorescein, 4',5'-dihydroxy | |
| C1=CC=C2C(=C1)C(=O)OC23C4=C(C(=C(C=C4)O)O)OC5=C3C=CC(=C5O)O | |
| 364.309 | |
| 364.31 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes