Learn More
Invitrogen™ Galectin 9 Monoclonal Antibody (9M1-3), eFluor™ 660, eBioscience™

Description
Description: This 9M1-3 monoclonal antibody reacts with human Galectin-9. Galectin-9 is one of fifteen members of the galectin family of glycan-binding proteins. It is expressed on activated T helper cells and Foxp3+ T regulatory cells and plays a critical role in the regulation of immune function. Interaction of Galectin-9 with its ligand, TIM3, induces apoptosis in Th1 and Th17, inhibits Th17 polarization and IFN gamma expression, and drives Treg expansion. Galectin-9 is also thought to contribute to the survival and expansion of some tumors by inhibiting the anti-tumor response and promoting immune evasion. Along with Galectin-1 and Galectin-3, high levels of Galectin-9 expression are associated with poor cancer prognosis, making these potential therapeutic targets. Applications Reported: This 9M1-3 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 9M1-3 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.25 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. eFluor® 660 is a replacement for Alexa Fluor® 647. eFluor® 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitati...
Specifications
Specifications
| Antigen | Galectin 9 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 9M1-3 |
| Concentration | 5 μL/Test |
| Conjugate | eFluor 660 |
| Formulation | PBS with BSA and 0.09% sodium azide; pH 7.2 |
| Gene | Lgals9 |
| Gene Accession No. | O00182 |
| Gene Alias | 36 kDa beta-galactoside-binding lectin; AA407335; AI194909; AI265545; ecalectin; Gal9; gal-9; galectin 9; Galectin9; galectin-9; HOM HD 21; HUAT; Lectin galactose binding soluble 9 (Galectin-9); lectin, galactose binding, soluble 5; lectin, galactose binding, soluble 9; Lectin, galactose binding, soluble 9 (Galectin-9); lectin, galactoside binding soluble 9; lectin, galactoside-binding, soluble, 9; LGALS35; Lgals5; LGALS9; LGALS9A; tumor antigen HOM-HD-21; UAT; urate transporter/channel; urate transporter/channel protein |
| Show More |