missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Galectin 2 Monoclonal Antibody (K70037_5D10), Invitrogen™

Catalog No. MA556270
Change view
Click to view available options
Quantity:
50 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA556270 200 μL
MA555890 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA556270 Supplier Invitrogen™ Supplier No. MA556270
Only null left

Mouse Monoclonal Antibody

Sequence of this protein is as follows: MTGELEVKNM DMKPGSTLKI TGSIADGTDG FVINLGQGTD KLNLHFNPRF SESTIVCNSL DGSNWGQEQR EDHLCFSPGS EVKFTVTFES DKFKVKLPDG HELTFPNRLG HSHLSYLSVR GGFNMSSFKL KE

The protein encoded by this gene is a soluble beta-galactoside binding lectin. The encoded protein is found as a homodimer and can bind to lymphotoxin-alpha. A single nucleotide polymorphism in an intron of this gene can alter the transcriptional level of the protein, with a resultant increased risk of myocardial infarction.

Specifications

Antigen Galectin 2
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone K70037_5D10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene LGALS2
Gene Accession No. P05162
Gene Alias 2200008F12Rik; AI324147; Beta-galactoside-binding lectin L-14-II; gal-2; galectin 2; galectin-2; HL14; lactose-binding lectin 2; lectin galactoside-binding soluble 2 (galectin 2); lectin, galactose-binding, soluble 2; lectin, galactoside binding soluble 2; lectin, galactoside-binding, soluble, 2; lectin, galactoside-binding, soluble, 2 (galectin 2); LEG2; Lgals2; S-Lac lectin 2
Gene Symbols LGALS2
Host Species Mouse
Immunogen Recombinant human LGALS2 (immunogen range is 1-132).
Purification Method Protein A
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3957
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less