missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Galectin 1 Recombinant Rabbit Monoclonal Antibody (23GB6280)
GREENER_CHOICE

Catalog No. MA553954
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA553954 100 μL
MA553955 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA553954 Supplier Invitrogen™ Supplier No. MA553954
Only null left

Rabbit Recombinant Monoclonal Antibody

Galectin 1 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

Galectin-1 belongs to a large family of carbohydrate-binding proteins called lectins. Galectin-1can be either monomeric or homodimeric and is found in a wide variety of cells and tissue types. Galectin-1 can control cell growth, proliferation, induce apoptosis of activated T cells while it can also modulate cytokine secretion or inhibit pro-inflammatory cytokine production. Galectin-1 plays an important role in acute and chronic inflammation.

Specifications

Antigen Galectin 1
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB6280
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene LGALS1
Gene Accession No. P09382, P11762, P16045
Gene Alias 14 kDa laminin-binding protein; 14 kDa lectin; AA410090; beta-galactoside binding protein; beta-galactoside-binding lectin; beta-galactoside-binding lectin L-14-I; beta-galactoside-binding protein 14kDa; DKFZp686E23103; GAL1; GAL-1; Galaptin; Galbp; galectin 1; galectin 1; LGALS1; Galectin1; galectin-1; galectin-1-like protein; GBP; HBL; H-Galectin-1; HLBP14; HPL; L14; L-14.5; Lactose-binding lectin 1; Lect14; Lectin galactoside-binding soluble 1; lectin, galactose binding, soluble 1; lectin, galactoside binding soluble 1; lectin, galactoside-binding, soluble, 1; lectin, galactoside-binding, soluble, 1 (galectin 1); Lgals1; LGAS1; OPG-1; pm12; Putative MAPK-activating protein PM12; RL 14.5; similar to galactose-binding lectin; S-Lac lectin 1; unnamed protein product
Gene Symbols LGALS1
Host Species Rabbit
Immunogen A synthesized peptide derived from human Galectin 1 (1-40AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16852, 3956, 56646
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less