missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Galactinol Hydrate 99.0+%, TCI America™
$221.83 - $1073.70
Chemical Identifiers
| CAS | 3687-64-7 |
|---|---|
| Molecular Formula | C12H26O13 |
| Molecular Weight (g/mol) | 378.327 |
| InChI Key | HGCURVXTXVAIIR-FMQVHOFPSA-N |
| Synonym | 1-O-alpha-D-Galactopyranosyl-L-myo-inositol |
| PubChem CID | 45052268 |
| IUPAC Name | (1S,2R,4S,5S)-6-[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxycyclohexane-1,2,3,4,5-pentol;dihydrate |
| SMILES | C(C1C(C(C(C(O1)OC2C(C(C(C(C2O)O)O)O)O)O)O)O)O.O.O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
G0298100MG
|
TCI America
G0298100MG |
100 mg |
Each for $221.83
|
|
|||||
|
G02981G
|
TCI America
G02981G |
1 g |
Each for $1,073.70
|
|
|||||
Chemical Identifiers
| 3687-64-7 | |
| 378.327 | |
| 1-O-alpha-D-Galactopyranosyl-L-myo-inositol | |
| (1S,2R,4S,5S)-6-[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxycyclohexane-1,2,3,4,5-pentol;dihydrate |
| C12H26O13 | |
| HGCURVXTXVAIIR-FMQVHOFPSA-N | |
| 45052268 | |
| C(C1C(C(C(C(O1)OC2C(C(C(C(C2O)O)O)O)O)O)O)O)O.O.O |
Specifications
| 3687-64-7 | |
| White | |
| 100 mg | |
| HGCURVXTXVAIIR-FMQVHOFPSA-N | |
| (1S,2R,4S,5S)-6-[(2R,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxycyclohexane-1,2,3,4,5-pentol;dihydrate | |
| 45052268 | |
| ≥99.0% (HPLC) | |
| Galactinol Hydrate |
| 221°C | |
| C12H26O13 | |
| 1-O-alpha-D-Galactopyranosyl-L-myo-inositol | |
| C(C1C(C(C(C(O1)OC2C(C(C(C(C2O)O)O)O)O)O)O)O)O.O.O | |
| 378.327 | |
| 342.30 | |
| Crystalline Powder |
Safety and Handling
TSCA : No