accessibility menu, dialog, popup

UserName

CAS RN 3687-64-7

HOOOOHHOHOOHOHOHOHOH

CAS RN 3687-64-7

IUPAC Name: 6-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}cyclohexane-1,2,3,4,5-pentol
Synonyms: 6-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}cyclohexane-1,2,3,4,5-pentol
Molecular Weight (g/mol): 342.3
Molecular Formula: C12H22O11
InChi Key: VCWMRQDBPZKXKG-UHFFFAOYNA-N
SMILES: OCC1OC(OC2C(O)C(O)C(O)C(O)C2O)C(O)C(O)C1O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results