missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium sputtering target, 76.2mm dia x 3.18mm thick

Catalog No. AA45929KS
Change view
Click to view available options
Quantity:
1 Ea.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA45929KS 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA45929KS Supplier Thermo Scientific Chemicals Supplier No. 045929KS

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 7440-54-2 | Gd | 157.25 g/mol

Gadolinium is use in neutron radiography and in shielding of nuclear reactors. Used in nuclear marine propulsion systems, used for decarboxylation of oxaloacetic acid, convertion of orto- to para-hydrogen and polymerization of ethylene. It is also used for making gadolinium yttrium, fabrication, color TV tubes, compact discs and computer memory.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7440-54-2
Molecular Formula Gd
Molecular Weight (g/mol) 157.25
MDL Number MFCD00011022
InChI Key UIWYJDYFSGRHKR-UHFFFAOYSA-N
Synonym atom, gadolinio, unii-au0v1lm3jt, trihydride, au0v1lm3jt, foil, 3n, gadolinium, elemental, ingot, gadolinium, chips, compounds
PubChem CID 23982
ChEBI CHEBI:33375
IUPAC Name gadolinium
SMILES [Gd]

Specifications

Quantity 1 Ea.
Solubility Information Soluble in magnesium and diluted acid. Insoluble in water.
Chemical Name or Material Gadolinium sputtering target, 76.2mm dia. x 3.18mm thick
EINECSNumber 231-162-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only