missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium(III) oxide, REacton™, 99.99% (REO)

Catalog No. p-4781901
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1129009 10 g
AA1129018 50 g
AA1129030 250 g
AA11290A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA1129009 Supplier Thermo Scientific Chemicals Supplier No. 01129009
Only null left

CAS: 12064-62-9 | Gd2O3 | 362.50 g/mol

Gadolinium(III) oxide is used as a raw material for various fluorescent compounds, absorption material in atomic reactions, nuclear fuels, magnetic bubble material and screen-sensitivity increasing material. It is also used in glass and electronic industries. It also acts as a neutron shield, a catalyst, a dielectric ceramic and is used for filament coatings, special glasses, lasers, masers and telecommunication. It also serves as a laboratory reagent. It plays an important role as a potential contrast agent for magnetic resonance imaging (MRI).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12064-62-9
Molecular Formula Gd2O3
Molecular Weight (g/mol) 362.50
MDL Number MFCD00011026
InChI Key CMIHHWBVHJVIGI-UHFFFAOYSA-N
Synonym gadolinia, gadolinium oxide, gadolinium iii oxide, gadolinium trioxide, digadolinium trioxide, gadolinium sesquioxide, gadolinium 3+ oxide, unii-5480d0nhlj, gadolinium 3+ ; oxygen 2-, digadolinium 3+ ion trioxidandiide
PubChem CID 159427
SMILES [O--].[O--].[O--].[Gd+3].[Gd+3]

Specifications

Melting Point 2330°C
Color White
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.99% (REO)
Solubility Information Soluble in acid. Insoluble in water.
IUPAC Name digadolinium(3+) trioxidandiide
Formula Weight 362.5
Density 7.407 g/cm3
Chemical Name or Material Gadolinium(III) oxide, Rare Earth Oxides Content
Hazard Category H319
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 235-060-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only