missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium(III) oxide, 99.9% (REO)

Catalog No. AA3635514
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3635514 25 g
AA3635522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3635514 Supplier Thermo Scientific Chemicals Supplier No. 03635514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 12064-62-9 | Gd2O3 | 362.50 g/mol

Gadolinium(III) oxide is used for vacuum deposition, synthesis of raw material for intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12064-62-9
Molecular Formula Gd2O3
Molecular Weight (g/mol) 362.50
MDL Number MFCD00011026
InChI Key CMIHHWBVHJVIGI-UHFFFAOYSA-N
Synonym gadolinia, gadolinium oxide, gadolinium iii oxide, gadolinium trioxide, digadolinium trioxide, gadolinium sesquioxide, gadolinium 3+ oxide, unii-5480d0nhlj, gadolinium 3+ ; oxygen 2-, digadolinium 3+ ion trioxidandiide
PubChem CID 159427
IUPAC Name gadolinium(3+);oxygen(2-)
SMILES [O--].[O--].[O--].[Gd+3].[Gd+3]

Specifications

Melting Point 2330°C
Physical Form Sintered Lump
Quantity 25 g
Merck Index 14,4326
Solubility Information Soluble in acids. Insoluble in water.
Formula Weight 362.5
Percent Purity 99.9%
Density 7.407 g/mL
Assay (REO)
Chemical Name or Material Gadolinium(III) oxide
Hazard Category H319
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 235-060-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only