missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium(III) fluoride, anhydrous, REacton™, 99.99% (REO)

Catalog No. AA1129309
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1129309 10 g
AA1129318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1129309 Supplier Thermo Scientific Chemicals Supplier No. 01129309

CAS: 13765-26-9 | F3Gd | 214.25 g/mol

Gadolinium(III) fluoride, anhydrous is employed in the synthesis of non-oxide glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 13765-26-9
Molecular Formula F3Gd
Molecular Weight (g/mol) 214.25
MDL Number MFCD00016113
InChI Key TYIZUJNEZNBXRS-UHFFFAOYSA-K
IUPAC Name gadolinium(3+) trifluoride
SMILES [F-].[F-].[F-].[Gd+3]

Specifications

Melting Point 1231°C
Color White
Physical Form Crystalline
Quantity 10 g
Solubility Information Insoluble in water.
Formula Weight 214.25
Percent Purity 99.99%
Grade Reacton
Odor Odorless
Sensitivity Hygroscopic
Density 7.1 g/mL
Assay (REO)
Chemical Name or Material Gadolinium(III) fluoride, Anhydrous
Show More Show Less
Hazard Category H314
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
EINECSNumber 237-369-4
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only