CAS RN 13765-26-9
IUPAC Name:
gadolinium(3+) trifluoride
Synonyms:
gadolinium(3+) trifluoride
Molecular Weight (g/mol):
214.25
Molecular Formula:
F3Gd
InChi Key:
TYIZUJNEZNBXRS-UHFFFAOYSA-K
SMILES:
[F-].[F-].[F-].[Gd+3]
1
–
3
of
3
results
1
Gadolinium(III) fluoride, anhydrous, REacton™, 99.9% (REO)
CAS: 13765-26-9 Molecular Formula: F3Gd Molecular Weight (g/mol): 214.25 MDL Number: MFCD00016113 InChI Key: TYIZUJNEZNBXRS-UHFFFAOYSA-K IUPAC Name: gadolinium(3+) trifluoride SMILES: [F-].[F-].[F-].[Gd+3]
2
Gadolinium(III) fluoride, anhydrous, REacton™, 99.99% (REO)
CAS: 13765-26-9 Molecular Formula: F3Gd Molecular Weight (g/mol): 214.25 MDL Number: MFCD00016113 InChI Key: TYIZUJNEZNBXRS-UHFFFAOYSA-K IUPAC Name: gadolinium(3+) trifluoride SMILES: [F-].[F-].[F-].[Gd+3]
3
Gadolinium(III) fluoride, anhydrous, 99.9% (REO)
CAS: 13765-26-9 Molecular Formula: F3Gd Molecular Weight (g/mol): 214.25 MDL Number: MFCD00016113 InChI Key: TYIZUJNEZNBXRS-UHFFFAOYSA-K IUPAC Name: gadolinium(3+) trifluoride SMILES: [F-].[F-].[F-].[Gd+3]