missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gadolinium(III) fluoride, anhydrous, 99.9% (REO)
Description
Gadolinium(III) fluoride, anhydrous is employed in the synthesis of non-oxide glasses.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 1231°C |
| Color | White |
| Physical Form | Pieces |
| Quantity | 50 g |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Density | 7.1 g/mL |
| Solubility Information | Insoluble in water. |
| Formula Weight | 214.25 |
| Percent Purity | 99.9% |
| Show More |
Safety and Handling
| Hazard Category | H314 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
| DOTInformation | Hazard Class: 8; Packaging Group: III |
| EINECSNumber | 237-369-4 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only