missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gadolinium(III) 2,4-pentanedionate hydrate, 99%, Thermo Scientific™
$123.93 - $123.93
Chemical Identifiers
| CAS | 64438-54-6 |
|---|---|
| Molecular Formula | C15H30GdO9 |
| Molecular Weight (g/mol) | 511.646 |
| MDL Number | MFCD00192188 |
| InChI Key | RFLUHCFSJOSWMI-FEUFEBADSA-N |
| Synonym | gadolinium iii acetylacetonate hydrate-gd reo |
| PubChem CID | 131675867 |
| IUPAC Name | gadolinium;(E)-4-hydroxypent-3-en-2-one;trihydrate |
| SMILES | CC(=CC(=O)C)O.CC(=CC(=O)C)O.CC(=CC(=O)C)O.O.O.O.[Gd] |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6038006
|
Thermo Scientific Chemicals
H6038006 |
5 g |
Each for $123.93
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 64438-54-6 | |
| 511.646 | |
| RFLUHCFSJOSWMI-FEUFEBADSA-N | |
| 131675867 | |
| CC(=CC(=O)C)O.CC(=CC(=O)C)O.CC(=CC(=O)C)O.O.O.O.[Gd] |
| C15H30GdO9 | |
| MFCD00192188 | |
| gadolinium iii acetylacetonate hydrate-gd reo | |
| gadolinium;(E)-4-hydroxypent-3-en-2-one;trihydrate |
Specifications
| 64438-54-6 | |
| C15H30GdO9 | |
| 5 g | |
| gadolinium iii acetylacetonate hydrate-gd reo | |
| CC(=CC(=O)C)O.CC(=CC(=O)C)O.CC(=CC(=O)C)O.O.O.O.[Gd] | |
| 511.646 | |
| 454.60 (Anhydrous) | |
| Gadolinium(III) 2,4-pentanedionate hydrate |
| 143°C | |
| MFCD00192188 | |
| Hygroscopic | |
| RFLUHCFSJOSWMI-FEUFEBADSA-N | |
| gadolinium;(E)-4-hydroxypent-3-en-2-one;trihydrate | |
| 131675867 | |
| 99% |
Safety and Handling
EINECSNumber : 238-186-2
TSCA : No
Recommended Storage : Ambient temperatures