missing translation for 'onlineSavingsMsg'
Learn More

Gadolinium(III) 2,4-pentanedionate hydrate, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H6038006

 View more versions of this product

Catalog No. AAH6038006


Only null left
Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Gadolinium(III) 2,4-pentanedionate hydrate
143°C
MFCD00192188
Hygroscopic
RFLUHCFSJOSWMI-FEUFEBADSA-N
gadolinium;(E)-4-hydroxypent-3-en-2-one;trihydrate
131675867
99%
64438-54-6
C15H30GdO9
5 g
gadolinium iii acetylacetonate hydrate-gd reo
CC(=CC(=O)C)O.CC(=CC(=O)C)O.CC(=CC(=O)C)O.O.O.O.[Gd]
511.646
454.60 (Anhydrous)

Chemical Identifiers

64438-54-6
511.646
RFLUHCFSJOSWMI-FEUFEBADSA-N
131675867
CC(=CC(=O)C)O.CC(=CC(=O)C)O.CC(=CC(=O)C)O.O.O.O.[Gd]
C15H30GdO9
MFCD00192188
gadolinium iii acetylacetonate hydrate-gd reo
gadolinium;(E)-4-hydroxypent-3-en-2-one;trihydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 238-186-2

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions