missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gabapentin 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 166°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 171.24 |
| Percent Purity | ≥98.0% (GC,T) |
| Grade | Research |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Gabapentin |
Safety and Handling
| RTECSNumber | GU6496000 |
| TSCA | No |