accessibility menu, dialog, popup

UserName

H2NOHO

CAS RN 60142-96-3

IUPAC Name: 2-[1-(aminomethyl)cyclohexyl]acetic acid
Synonyms: gabapentin gabapentin preparation gabapentin GR aventis brand of gabapentin gabapentino gabapentine gabapentin (product) parke davis brand of gabapentin apotex brand of gabapentin gabapentin (substance)
Molecular Weight (g/mol): 171.24
Molecular Formula: C9H17NO2
InChi Key: UGJMXCAKCUNAIE-UHFFFAOYSA-N
SMILES: NCC1(CC(O)=O)CCCCC1
Quantity 
  • (1)
  • (2)
  • (2)
Grade 
  • (1)
  • (2)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (2)