missing translation for 'onlineSavingsMsg'
Learn More

Gabapentin 98.0+%, TCI America™

Supplier:  TCI America G031825G

Quantity 25 g

 View more versions of this product

Catalog No. G031825G


Only null left

Description

Description

Specifications

Chemical Identifiers

60142-96-3
171.24
UGJMXCAKCUNAIE-UHFFFAOYSA-N
3446
2-[1-(aminomethyl)cyclohexyl]acetic acid
C9H17NO2
MFCD00865286
gabapentin, neurontin, gabapentine, 1-aminomethyl cyclohexaneacetic acid, 2-1-aminomethyl cyclohexyl acetic acid, aclonium, gabapentinum, gabapetin, gabapentino, fanatrex
CHEBI:42797
C1CCC(CC1)(CC(=O)O)CN

Specifications

166°C
60142-96-3
C9H17NO2
gabapentin, neurontin, gabapentine, 1-aminomethyl cyclohexaneacetic acid, 2-1-aminomethyl cyclohexyl acetic acid, aclonium, gabapentinum, gabapetin, gabapentino, fanatrex
C1CCC(CC1)(CC(=O)O)CN
25 g
3446
171.24
Crystalline Powder
Gabapentin
White
MFCD00865286
UGJMXCAKCUNAIE-UHFFFAOYSA-N
2-[1-(aminomethyl)cyclohexyl]acetic acid
171.24
CHEBI:42797
≥98.0% (GC,T)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : GU6496000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions