missing translation for 'onlineSavingsMsg'
Learn More

Gabapentin Related Compound E Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015148
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015148 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015148 Supplier Merck Emd Millipore Supplier No. PHR201350MG
Only null left

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Specifications

CAS 67950-95-2
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C9H14O4
MDL Number MFCD04037521
Synonym 1-Carboxycyclohexaneacetic acid; CARBOXYMETHYL-CYCLOHEXANECARBOXYLIC ACID
Type Certified Reference Material
Quantity 50 mg
Formula Weight 186.21
Grade Certified Reference Material